Learn More
Electron Microscopy Sciences Gilder Rectangular Slotted Grid G100/400, Copper
Supplier:  Electron Microscopy Sciences G1040CU
Encompass_Preferred
A reliable support specimen grid source. Features: well-defined grid bars, maximum open area, and a matt/shiny side. Each grid is individually inspected. 3.05 mm diameter, 0.7 mil (18um) thickness.
Material: Copper
Pitch: 250/62 µm
Hole: 205/37 µm
Bar: 45/25 µm
Catalog No. 50-283-24
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.