Learn More
Electron Microscopy Sciences Veco Grid L0.2 x 0.5, Copper with 200 x 500 µm Rectangle Slot
Supplier: Electron Microscopy Sciences 0502CU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Copper
Slot: 200 x 500 µm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.