Learn More
Ace Glass, Inc. REACTOR BODY PLAIN 2L
Supplier: Ace Glass, Inc. 643716
2000mL one-piece pressure vessel body featuring a #25 Ace-Thred center neck, three #15 and one #7 side necks. Vessel also features a 0-8mm, 28/15 ball joint bottom outlet.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.