Learn More
Ace Glass, Inc. 1000mL, two-piece pressure reactor complete with 0-8mm valve and 28/15 ball joint bottom outlet, 100mm head with #15center, (3)#15, (1)#7 side necks, addition funnel, condenser, pressure manifold, stirring & temperature control assemblies
Supplier: Ace Glass, Inc. 6425211
1000mL, two-piece pressure reactor complete with 0-8mm valve and 28/15 ball joint bottom outlet, 100mm head with #15center, (3)#15, (1)#7 side necks, addition funnel, condenser, pressure manifold, stirring & temperature control assemblies
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.