Learn More
Ace Glass, Inc. 500mL, one-piece jacketed pressure reactor with #25 center, (3)#15, (1)#7 side necks, 0-8mm valve and 28/15 ball joint bottom outlet, #11 Ace-Thred inlet/outlet, complete with addition funnel, condenser, manifold & stirring assemblies
Supplier:  Ace Glass, Inc. 6439343
Encompass_Preferred
Complete, one-piece 500mL jacketed pressure reactor is designed to perform low to moderate positive pressure reactions, synthesis, and catalysis. Flask has a #25 center neck, (3) #15 side necks, and (1) #7 side neck. Flask bottom outlet is 0-8mm 28/15 ball joint. Jacket inlet/outlet has a #11 Ace-Thred for use with 3/8 I.D. tubing. Rated 45 PSI at 100 degrees C. Reactor system includes addition funnel, condenser, manifold, and stirring assemblies.
Catalog No. 50-972-967
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.