Learn More
Electron Microscopy Sciences Reagent Alcohol 50% 500 ML
Supplier:  Electron Microscopy Sciences 2620602
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Used in Alizarin Red S for calcium deposits staining. See references: McGee-Russell, S.M., J. Histochem., 6:22-42, 1958 or Clark, G., (ed), Staining Procedures, 3rd d., Williams & Wilkins, Baltimore, p. 140, c. 1973.
Catalog No. 50-317-36
missing translation for 'QuantityLeft'