Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reagent Alcohol, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, anhydrous, SDA 3A, 90 ±1%, Honeywell™

Catalog No. p-7112043 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
19 L
4 x 4L
Packaging:
Metal Can
Poly Bottle

Chemical Identifiers

CAS 64-17-5, 67-56-1, 67-63-0
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

CAS Max % 90.0000%, 5.0000%, 5.0000%
Density 0.7847 g/cm3
Boiling Point 78.32°C
Quantity 19 L
Flash Point 15°C
Packaging Metal Can
Assay Percent Range 94 to 96%
UN Number UN1987
Beilstein 1718733
Water 0.5% max.
Solubility Information Miscible in Water at 20°C
Grade ACS Reagent
Chemical Name or Material Reagent Alcohol, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, anhydrous, SDA 3A, 90 ±1%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Alcohols, n.o.s.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.