Learn More
Thermo Scientific™ Reagents for Free Chlorine
Keep your Thermo Scientific™ Orion™ Chlorine XP chlorine analyzer well supplied with the reagents it needs for Free Chlorine and Total Chlorine measurement (DPD method).
Supplier:  Thermo Scientific™ CXPRGDPD1F
 View more versions of this product
Catalog No. 13-642-104
Description
Description
Keep your chlorine analyzer well supplied with the reagents it needs for Free Chlorine and Total Chlorine measurement. The Chlorine XP continuously monitors a process stream using an automated DPD test for free and optionally totalchlorine. For use with the Orion™ Chlorine XP.
Specifications
Specifications
5 Sets of DPD1 reagents for Free Chlorine
Reagent
600 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.