Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Adenosine Deaminase 2/CECR1 Protein
Shop All R&D Systems Products
Click to view available options
:
10ug; Carrier Free
Description
The Recombinant Human Adenosine Deaminase 2/CECR1 Protein is derived from CHO. The Recombinant Human Adenosine Deaminase 2/CECR1 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Formulation | Supplied as a 0.2μm filtered solution in HEPES, NaCl, Glycerol. |
Gene ID (Entrez) | 51816 |
Molecular Weight (g/mol) | 57 kDa |
Name | Adenosine Deaminase 2/CECR1 |
Quantity | 10μg |
Source | Chinese Hamster Ovary cell line,CHO-derived human Adenosine Deaminase 2/CECR1 protein Ile30-Lys511,with a C-terminal 6-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Purity or Quality Grade | >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction