Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Adenosine Deaminase/ADA Protein
Shop All R&D Systems Products
Click to view available options
Quantity:
10 μg
Description
The Recombinant Human Adenosine Deaminase/ADA Protein is derived from Sf 21 (baculovirus). The Recombinant Human Adenosine Deaminase/ADA Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Formulation | Supplied as a 0.2μm filtered solution in MOPS, NaCl and Tween™ 80. |
Gene ID (Entrez) | 100 |
Molecular Weight (g/mol) | 42 kDa |
Name | Adenosine Deaminase/ADA |
Quantity | 10 μg |
Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Adenosine Deaminase/ADA protein Met1-Leu363,with a C-terminal 6-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
Conjugate | Unconjugated |
Recombinant | Recombinant |
Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction