Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human VEGF 165 Protein

Catalog No. BTVEGF050 Shop All R&D Systems Products
Click to view available options
Quantity:
This item is not returnable. View return policy
This item is not returnable. View return policy
Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. Conn, G. et al. (1990) Proc. Natl. Acad. Sci. USA 87:1323. The ED50 for this effect is 1.50-12.0 ng/mL.

Specifications

Accession Number

NP_001165097.1

For Use With (Application)

Bioactivity

Formulation

Lyophilized from a 0.2 μm filtered solution in Sodium Acetate.

Gene ID (Entrez)

7422

Molecular Weight (g/mol)

M.W. Observed: 19-21 kDa, under reducing conditions., M.W. Theoretical: 19.2 kDa (monomer)

Quantity

50 μg

Source

E. coli-derived human VEGF protein Ala27-Arg191

Storage Requirements

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.

Endotoxin Concentration

<0.01 EU per 1 μg of the protein by the LAL method.

Gene Alias

MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor

Conjugate

Unconjugated

Reconstitution

Reconstitute the 10 μg size at 100 μg/mL in sterile deionized water. Reconstitute all other sizes at 500 μg/mL in sterile deionized water.

Purity or Quality Grade

>97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie® Blue Staining.

Protein

VEGF

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.