Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-17E Protein
Shop All R&D Systems Products
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Molecular Weight (g/mol):
17.6 kDa
17.6 kDa (monomer)
Description
The Recombinant Mouse IL-17E Protein is derived from E. coli. The Recombinant Mouse IL-17E Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
Gene ID (Entrez) | 140806 |
Molecular Weight (g/mol) | 17.6 kDa (monomer) |
Name | IL-17E/IL-25 |
Quantity | 25 μg |
Source | E. coli-derived mouse IL-17E/IL-25 protein Val17-Ala169,with and without an N-terminal Met |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Structural Form | Disulfide-linked homodimer |
Endotoxin Concentration | <0.01 EU per 1μg of the protein by the LAL method. |
Conjugate | Unconjugated |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction