Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Mouse TGF-beta RI/ALK-5 Fc Chimera Recombinant Protein
Shop All R&D Systems Products
Click to view available options
:
50ug; Carrier Free
Description
The Recombinant Mouse TGF-beta RI/ALK-5 Fc Chimera Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse TGF-beta RI/ALK-5 Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
Gene ID (Entrez) | 21812 |
Molecular Weight (g/mol) | 37.5 kDa (monomer) |
Name | TGF-beta RI/ALK-5 Fc Chimera |
Quantity | 50 μg |
Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse TGF-beta RI/ALK-5 protein, (N-terminus) Mouse TGF-beta RI (Ala21-Glu121) & (Thr22-Glu121) Accession &Num; BAA05023 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Structural Form | Disulfide-linked homodimer |
Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
Conjugate | Unconjugated |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction