Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Viral HPV E6 Protein

Catalog No. AP120025 Shop All R&D Systems Products
Click to view available options
Quantity:
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Viral HPV E6 Protein is derived from E. coli. The Recombinant Viral HPV E6 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number

P03126.1

Formulation

Supplied as a 0.2 μm filtered solution in HEPES, NaCl, DTT.

Gene ID (Entrez)

1489078

Name

HPV E6

Quantity

25 μg

Source

E. coli-derived viral E6 protein Met1 - Leu158 with a N-terminal Ser-His Contains Cys to Ser substitutions at the following positions: 23, 58, 87, 104, 118, 147

Storage Requirements

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.

Conjugate

Unconjugated

Recombinant

Recombinant

Purity or Quality Grade

>98%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.