Learn More
Omnimed Inc Acrylic Refrigerator Lock Box with Combo Lock (4.25"H X 12"W X 6"D)
Designed for the storage of insulin or other refrigerated medicines, keep your medicine safe and secure. Each box is keyed using a combo lock. Boxes have pre-drilled holes for mounting to the underside of wire, glass shelves or cabinets.