Learn More
ACROBiosystems ACROBiosystems Human Reg4 Protein, His Tag
Supplier:  ACROBiosystems RE4H52211MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Recombinant Protein;1MG;Human Reg4, His Tag (RE4-H5221) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Pro 158 (Accession # AAH17089).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.7 kDa. The protein migrates as 18 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6928
missing translation for 'QuantityLeft'