Learn More
Chemglass Life Sciences Replacement Stopcock for Chemglass Polycarbonate Desiccator (STOPCOCK ONLY)
SDP
Supplier:  Chemglass Life Sciences CG122613
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Replacement part for CG1226 series desiccators. The supplied polypropylene stopcock has a PTFE plug which turns easily, allowing a slow, gentle entrance of air to prevent unnecessary turbulence and crucible damage. Stopcock port accepts 1/4” I.D. tubing.
Catalog No. CG122613
missing translation for 'QuantityLeft'