Learn More
Hach Company Replacement DO sensor for Sension+ 5049 multisensor
Supplier:  Hach Company LZW9173.99
Encompass_Preferred
Laboratory and field probes for use with Sension+ meters.A wide selection of Sension+ electrodes are available to meet almost all laboratory and field applications. Measureable parameters include pH, Dissolved Oxygen, Conductivity, ORP, Ammonia, Ammonium, Chloride, Fluoride, Nitrate, and Sodium. All pH probes are delivered with storage tubes. Field probes are equipped with screwable calibration tubes, serving excellent usability in the field and minimising buffer consumption. Several multi probes are available for Sension+ portable multi meters, saving time and money due to the simultaneous measurement of up to three parameters, combined with convenient handling.
Catalog No. 50-199-8000
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.