Learn More
Honeywell BW™ Replacement MICROpeL Combustible (LEL) Sensor
Honeywell BW™ Replacement MICROpeL Combustible (LEL) Sensor for use with GasalertMicroClip gas detectors
Supplier: Honeywell BW™ SRWMP75C
Description
- 0 to 100% LEL, 0 to 5% v/v measuring range
- 1 and 0.1% default resolution
Specifications
GasAlertMicroClip and GasAlertMax XT II | |
Sensor | |
2.54 cm | |
Combustible (LEL) Sensor |