Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trajan Scientific and Medical™ Replacement Needle and Plunger Kit for PerkinElmer Autosampler Syringes

Catalog No. 033765 Shop All SGE Inc. Products
Click to view available options
Diameter (Metric) Outer Needle:
Needle Gauge:

Repair kit for PerkinElmer Autosystem™ and Clarus™ Removable Needle Autosampler Syringes

Includes:

  • 70mm cone tipped needle
  • 0.5μL volume

Choose Needle:

  • 26 gauge, 0.47 OD
  • 23 gauge, 0.63 OD

Specifications

Needle Gauge

23 G

Diameter (Metric) Outer Needle

0.63 mm

Product Type

Replacement Needle and Plunger Kit

Volume (Metric)

0.5 μL

Length (Metric) Needle

70 mm

Needle Point Style

Cone

For Use With (Equipment)

AutoSystem and Clarus Removable Needle Syringe

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.