Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trajan Scientific and Medical™ Replacement Needles for Agilent Plunger-in-Needle Syringes

Catalog No. 034715 Shop All SGE Inc. Products
Click to view available options
Volume (Metric):
Needle Gauge:

Quality needles to fit a SGE syringe in a range of lengths and tip styles.

  • Needle length is the exposed length of needle
  • Choose a different replacement needle by selecting syringe volume, needle length, OD and tip style

Specifications

Needle Gauge

23 G

Diameter (Metric) Inner Needle

0.22 mm

Diameter (Metric) Outer Needle

0.63 mm

Volume (Metric)

1 μL

Length (Metric) Needle

42 mm

Needle Point Style

Cone

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.