Learn More
Glass Expansion Replacement Captive Screws M4 for Brass Skimmer Adaptor Base for Agilent 7900 X Lens
Supplier:  Glass Expansion 708032061
Encompass_Preferred
Replacement Captive Screws M4 for Brass Skimmer Adaptor Base for Agilent 7900 X Lens. Package of 3 replacement M4 screws for the Agilent 7900 X Lens Brass Skimmer Adaptor Base (AT7905X-BR). These screws should be replaced if they are corroded or worn.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-3020
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.