Learn More
Med Vet International Rescue 1 Gallon Concentrate Disinfectant Cleaner and Deodorizer
Supplier:  Med Vet International PHVRESCUEGAL
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Rescue is a one-step disinfectant cleaner and deodorizer formulated with AHP (Accelerated Hydrogen Peroxide) for veterinary use. When used as directed, this product is highly effective against a wide variety of pathogenic microorganisms including bacteria, antibiotic resistant bacteria, viruses, fungi, mold and mildew including: Canine Parvovirus Canine Distemper Virus Enterococcus Microsporum Canis Streptococcus Trichophyton (Ringworm)
Catalog No. 50-209-2658
missing translation for 'QuantityLeft'