Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Resorufin 95.0+%, TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Color | Black-Green |
Quantity | 1 g |
Formula Weight | 213.19 |
Percent Purity | ≥95.0% (HPLC) |
Physical Form | Crystalline Powder |
Chemical Name or Material | Resorufin |
Safety and Handling
RTECSNumber | SP7698000 |
TSCA | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction