Learn More
Restek™ Replacement Inlet Cross-Disk Seals for Agilent 6890 GCs

Ideal for high-flow split applications >500mL/min. on Agilent 5890 GCs.
Supplier: Restek™ 20476
Description
- Siltek treatment provides enhanced inertness versus stainless steel
- All seals include washers
- I.D.: 0.8mm
Specifications
Seal | |
10 Pack | |
Gold-plated |
0.8 mm | |
Agilent 6890 GCs |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.