Learn More
AdipoGen Reticulol
SDP
Supplier:  AdipoGen BVT0011M001
Encompass_Preferred
Chemical. CAS 26246-41-3. Formula C11H10O5. MW 222.2. Isolated from Streptomyces sp. Antibiotic. Cyclic adenosine 3,5-monophosphate phosphodiesterase and topoisomerase I Topo I inhibitor. Antitumor compound.
Catalog No. 50-256-2150
missing translation for 'QuantityLeft'
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.