Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Retinol Acetate, Water Soluble, MP Biomedicals™

Catalog No. MP219931105 missing translation for 'brandsLinkText'
missing translation for 'orderingAttributeHoverText'
Quantity:

Often used in cell culture applications. Retinol Acetate, Water Soluble, MP Biomedicals (aka Vitamin A) is found in classical media formulations, such as M-199; NCTC-109 and William’s Medium E. Typical working concentration is ca. 3.44mg of retinol acetat

Vitamin A is a component of serum used to supplement classical media. Consequently, most serum-containing cell culture systems receive some level of vitamin A. The oxidized form of retinol is retinoic acid, which has hormone-like growth factor and differentiation activity for epithelial and related cell types, especially neural cells. Retinoic acid induces embryonic stem cells to differentiate towards neural cells.

Chemical Identifiers

CAS

127-47-9

Molecular Formula

C22H32O2

Molecular Weight (g/mol)

328.496

InChI Key

QGNJRVVDBSJHIZ-QHLGVNSISA-N

Synonym

retinyl acetate, vitamin a acetate, retinol acetate, retinol, acetate, all-trans-retinyl acetate, crystalets, vitamin a1 acetate, all-trans-retinol acetate, vitamin a alcohol acetate, davitan a 650

PubChem CID

638034

ChEBI

CHEBI:32095

IUPAC Name

[(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate

SMILES

CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCOC(=O)C)C)C

Specifications

Melting Point

57°C to 58°C (literature)

Quantity

5mg

Solubility Information

Soluble in chloroform (50mg/mL, clear yellow solution); absolute ethanol: soluble 25mg/mL (Lit.)

Formula Weight

328.5g/mol

Physical Form

Off-White Powder

Recommended Storage

Store at 0°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.