Learn More
NEOGEN Reveal 2.0 for E. coli O157:H7
SDP
Supplier:  NEOGEN 700002805
Encompass_Preferred
The Reveal 2.0 for E. coli O157:H7 test provides for the rapid recovery of E. coli O157:H7 and E. coli O157:NM organisms in beef matrices facilitating detection and presumptive identification of the target organisms in as little as 12 hours following enrichment.Approvals: AOAC-RI 011103Limit of Detection: 1 cfu/analytical unitTesting Time: 10 MinutesFor more information visit https://www.neogen.com/solutions/microbiology/reveal-2-e-coli-o157-h7
Catalog No. 50-201-5126
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.