Learn More
NEOGEN Reveal 2.0 for Group D1 Salmonella
The Reveal 2.0 for Group D1 Salmonella (including SE) Test System provides for the rapid recovery of Group D1 Salmonella spp. serovars (including S. Enteritidis) in layer or broiler house environmental samples.Approvals: AOAC 041602, NPIPLimit of Detection: 1 cfu/analytical unitTesting Time: 10 MinutesTime to Result: 48 HoursFor more information visit https://www.neogen.com/solutions/microbiology/reveal-2-group-d1-salmonella