Learn More
NEOGEN Reveal for Multi-Treenut
SDP
Supplier:  NEOGEN 700002602
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Reveal for Multi-Treenut Test is intended for the qualitative analysis of treenut (almond, cashew, hazelnut, pecan, pistachio, and walnut) residue in clean-in-place rinses and on environmental surfaces.Testing Time: 10 MinutesFor more information visit https://www.neogen.com/solutions/Allergens/reveal-multi-treenut
Catalog No. 50-201-5120