Learn More
Electron Microscopy Sciences Revolving Stand Screwdriver Set
Supplier:  Electron Microscopy Sciences 6207800
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Precision European quality screwdrivers in 9 sizes: .60; .80; 1.00; 1.20; 1.40; 1.60; 2.00; 2.50 and 3.00 mm. With hexagonal swivel heads to prevent rolling and knurled handles for a firm grip. Color identifier bands allow for easy selection and set screws hold the blades securely. Set contains 19 extra blades and 2 extra set screws. The set comes with a handsome revolving stand.
Catalog No. 50-996-512
missing translation for 'QuantityLeft'