Learn More
Medchemexpress LLC HY-111498 2mg Medchemexpress, RGX-104 CAS:610318-03-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1114982MG
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Medchemexpress, HY-111498 2mg RGX-104 CAS:610318-03-1 RGX-104 is a small-molecule LXR agonist that modulates innate immunity via transcriptional activation of the ApoE gene. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8887
missing translation for 'QuantityLeft'