Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RHC 80267, Tocris Bioscience™

Catalog No. p-4784404 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Diacylglycerol lipase inhibitor

Inhibitor of diacylglycerol lipase (IC50 values are 1.1 and 4 μM in rat cardiac myocytes and canine platelets respectively). Weakly inhibits phospholipases C and A2. Potentiates acetylcholine evoked relaxation in mesenteric arteries by the inhibition of cholinesterase activity (IC50 = 4 μM).

Chemical Identifiers

CAS 83654-05-1
Molecular Formula C20H34N4O4
Molecular Weight (g/mol) 394.52
MDL Number MFCD00210844
InChI Key RXSVYGIGWRDVQC-UHFFFAOYSA-N
Synonym 1,6-bis cyclohexyloximinocarbonyl hexane, 1,6-di o-carbamoyl cyclohexanone oxime hexane, 1,6-bis cyclohexyloximinocarbonylamino hexane, cyclohexanone, o,o'-1,6-hexanediylbis iminocarbonyl dioxime, quinolyl derivatives, d05lxg, cyclohexanone, o,o'-1,6-hexanediylbis iminocarbonyl oxime, rhc hplc , solid, o,o'-1,6-hexanediylbis iminocarbonyl dioxime cyclohexanone, cyclohexylideneamino n-6-cyclohexylideneamino oxycarbonylamino hexyl carbamate
PubChem CID 5063
IUPAC Name cyclohexylideneamino N-[6-({[(cyclohexylideneamino)oxy]carbonyl}amino)hexyl]carbamate
SMILES O=C(NCCCCCCNC(=O)ON=C1CCCCC1)ON=C1CCCCC1

Specifications

Quantity 10 mg
Formula Weight 394.51
Percent Purity >95%
Chemical Name or Material RHC 80267
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.