Learn More
MilliporeSigma™ Supelco™ RheBuild™ Kit
Kits include all of the tools and parts you need to repair your SupelPRO unit. The parts included are those most likely to be damaged, worn, or lost. Parts may vary, depending on valve model.
Supplier:  MilliporeSigma™ Supelco™ 54396U
 View more versions of this product
Catalog No. 11-100-9237
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
Kits for front-loading injectors include: rotor seal, stator face assembly, isolation seal, needle guide, needle port cleaner, 2 hex keys, operating instructions, and a mini-manual. Parts may vary, depending on valve model.
Specifications
Specifications
Liquid Chromatography
SupelPRO, 2-Position/10-Port
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
RheBuild is a registered trademark of IDEX Health & Science, LLC