Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rhodium black, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog No. p-4824907
Click to view available options
Quantity:
0.5 g
2 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-16-6
Molecular Formula Rh
Molecular Weight (g/mol) 102.91
MDL Number MFCD00011201 MFCD03458392
InChI Key MHOVAHRLVXNVSD-UHFFFAOYSA-N
Synonym fume, on carbon, rhodium, elemental, rodio, black, rhodium, metal, unii-dmk383dsac, on alumina, rh/al2o3, dmk383dsac
PubChem CID 23948
ChEBI CHEBI:33359
IUPAC Name rhodium
SMILES [Rh]

Specifications

Color Gray
Appearance gray powder
Assay Percent Range 99.9% (Metals basis)
Quantity 0.5 g
Odor Odorless
Form Powder
Chemical Name or Material Rhodium black
Hazard Category H228
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation DOT Class: 4.1, Packaging Group: III
EINECSNumber 231-125-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.