Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Risperidone, Tocris Bioscience™

Catalog No. p-4784406 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

5-HT2A antagonist

Risperidone is an atypical antipsychotic agent that displays 5-HT2A receptor antagonism. Also displays high affinity at D2 receptors (Ki values are 0.4 and 3.13 nM for 5-HT2A and D2 receptors respectively).

Chemical Identifiers

CAS 106266-06-2
Molecular Formula C23H27FN4O2
Molecular Weight (g/mol) 410.493
InChI Key RAPZEAPATHNIPO-UHFFFAOYSA-N
Synonym risperidone, risperidal, risperdal, risperdal consta, rispolept, risperin, rispolin, sequinan, risperidona, risperidonum
PubChem CID 5073
ChEBI CHEBI:8871
IUPAC Name 3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
SMILES CC1=C(C(=O)N2CCCCC2=N1)CCN3CCC(CC3)C4=NOC5=C4C=CC(=C5)F

Specifications

Quantity 10 mg
Formula Weight 410.48
Percent Purity >99%
Chemical Name or Material Risperidone
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.