Learn More
Medchemexpress LLC HY-10791 5mg Medchemexpress, Ritanserin CAS:87051-43-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY107915MG
Encompass_Preferred
Medchemexpress, HY-10791 5mg Ritanserin CAS:87051-43-2 Ritanserin (R 55667) is a highly potent, relatively selective, orally active, long acting antagonist of 5-HT 2 receptor , with an IC 50 of 0.9 nM, less active on Histamine H 1 , Dopamine D 2 , Adrenergic α 1 , Adrenergic α 2 receptors [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2305
missing translation for 'QuantityLeft'
Explore available promotions
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.