Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ritanserin, Tocris Bioscience™

Catalog No. p-4784410 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent 5-HT2 antagonist

Potent and long-acting 5-HT2 receptor antagonist (Ki = 0.39 nM). Anxiolytic in vivo.

Chemical Identifiers

CAS 87051-43-2
Molecular Formula C27H25F2N3OS
Molecular Weight (g/mol) 477.574
InChI Key JUQLTPCYUFPYKE-UHFFFAOYSA-N
Synonym ritanserin, tiserton, ritanserine, ritanserina, ritanserinum, ritanserine french, ritanserinum latin, ritanserina spanish, unii-145tfv465s, 6-2-4-bis 4-fluorophenyl methylene piperidin-1-yl ethyl-7-methyl-5h-thiazolo 3,2-a pyrimidin-5-one
PubChem CID 5074
ChEBI CHEBI:64195
IUPAC Name 6-[2-[4-[bis(4-fluorophenyl)methylidene]piperidin-1-yl]ethyl]-7-methyl-[1,3]thiazolo[3,2-a]pyrimidin-5-one
SMILES CC1=C(C(=O)N2C=CSC2=N1)CCN3CCC(=C(C4=CC=C(C=C4)F)C5=CC=C(C=C5)F)CC3

Specifications

Quantity 10 mg
Formula Weight 477.57
Percent Purity >99%
Chemical Name or Material Ritanserin
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.