Learn More
RNA Polymerase II, Polyclonal, Bethyl Laboratories

Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300653AM

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes the largest subunit of RNA polymerase II, the polymerase responsible for synthesizing messenger RNA in eukaryotes. The product of this gene contains a carboxy terminal domain composed of heptapeptide repeats that are essential for polymerase activity. These repeats contain serine and threonine residues that are phosphorylated in actively transcribing RNA polymerase. In addition, this subunit, in combination with several other polymerase subunits, forms the DNA binding domain of the polymerase, a groove in which the DNA template is transcribed into RNA.Specifications
RNA Polymerase II | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P08775, P24928 | |
POLR2A | |
C-terminal repeat | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
POLR2A | |
RNA POL2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20020, 5430 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.