Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ro 08-2750, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Description
Non-peptide inhibitor of NGF that binds the NGF dimer (KD ∽ 1 μM) possibly causing a conformational change. Selectively inhibits binding of NGF to p75NTR at submicromolar concentrations, and to both p75NTR and TrkA at concentrations > 5 μM. Prevents NGF-induced apoptosis in SK-N-MC cells. Suppresses cancer cell proliferation via regulation of Hippo signaling aand YAP phosphorylation.
Specifications
Specifications
Quantity | 1 mg |
Formula Weight | 270.24 |
Percent Purity | >96% |
Chemical Name or Material | Ro 08-2750 |
Safety and Handling
Recommended Storage | Store at -20°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction