Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ro 15-4513, Tocris Bioscience™

Catalog No. p-4784422 Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Benzodiazepine partial inverse agonist

High affinity benzodiazepine ligand. Ki values are 3.1 and 5.3 nM for diazepam-insensitive (DI) and diazepam-sensitive (DS) benzodiazepine receptors respectively. Acts as partial inverse agonist at recombinant DS α1-, α2-, α3- and α5-GABAA receptors. Displays partial agonism at DI α4- and α6-GABAA receptors. Antagonizes several behavioral and neurochemical effects of ethanol. Proconvulsant and anxiogenic.

Chemical Identifiers

CAS 91917-65-6
Molecular Formula C15H14N6O3
Molecular Weight (g/mol) 326.316
InChI Key CFSOJZTUTOQNIA-UHFFFAOYSA-N
Synonym unii-s5xgl82o5y, s5xgl82o5y, ethyl-8-azido-5,6-dihydro-5-methyl-6-oxo-4h-imidazo-1,4-benzodiazepine-3-carboxylate, ethyl 8-azido-5-methyl-6-oxo-4h-imidazo 1,5-a 1,4 benzodiazepine-3-carboxylate, ethyl 8-azido-6-dihydro-5-methyl-6-oxo-4h-imidazo 1,5-a 1,4 benzodiazepine-3-carboxylate, 4h-imidazo 1,5-a 1,4 benzodiazepine-3-carboxylic acid, 8-azido-5,6-dihydro-5-methyl-6-oxo-, ethyl ester, ethyl 12-azido-8-methyl-9-oxo-2,4,8-triazatricyclo 8.4.0.0^ 2,6 tetradeca-1 10 ,3,5,11,13-pentaene-5-carboxylate, d04ovj, d0m4cp, chembl6597
PubChem CID 5081
IUPAC Name ethyl 8-azido-5-methyl-6-oxo-4H-imidazo[1,5-a][1,4]benzodiazepine-3-carboxylate
SMILES CCOC(=O)C1=C2CN(C(=O)C3=C(N2C=N1)C=CC(=C3)N=[N+]=[N-])C

Specifications

Quantity 10 mg
Formula Weight 326.31
Percent Purity >98%
Chemical Name or Material Ro 15-4513
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.