Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ro 61-8048, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Ro 61-8048 is a potent and competitive kynurenine 3-monooxygenase (kynurenine 3-hydroxylase; KMO) inhibitor (Ki = 4.8 nM, IC50 = 37 nM). Increases kynurenic acid levels to concentrations that antagonize the glycine site of NMDA receptors. Brain penetrant and exhibits antidystonic, anticonvulsant and neuroprotective activities. Ro 61-8048 decreases nicotine self-administration in vivo. Ro 61-8048 prevents post-operative brain edema and consequent neuronal apoptosis in a rat model of surgically induced brain injury.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 421.45 |
Percent Purity | >99% |
Chemical Name or Material | Ro 61-8048 |
Safety and Handling
Recommended Storage | Store at +4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction