Learn More
Stoelting™ Robot Stereotaxic Manipulator Arm
Provides automated movement in all 3 orthogonal axes. Movement is integrated, controlled and directed by innovative software that provides a virtual 3-D map of the brain atlas with real time movement of the manipulators and visual marker of a probe's path through the brain
$28200.00 - $32150.00
Specifications
For Use With (Application) | Neuroscience |
---|---|
Size | 3 axes |
Animal | Mouse, Rat |
Catalog Number | Mfr. No. | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Model | Price | Quantity & Availability | |||||
10-000-189
![]() |
Stoelting™
51704 |
Add-On, Left |
Each for $28,200.00
|
|
|||||
10-000-191
![]() |
Stoelting™
51706 |
Add-on, Right |
Each for $32,150.00
|
|
|||||
Description
Designed to adapt conventional stereotaxic systems. Integrated with Paxinos and Watson's Rat Brain in Stereotaxic Coordinates, for either rat or mouse, the intuitive movement control enables accuracy and high throughput in all stereotaxic applications.
Software coordinates motor movement of all 3 axes (anterior-posterior, medial-lateral, and dorsal-ventral) probe placement in relation to 3-dimensional visual representation of a rat
Features:
- Easy calibration of frame coordinates
- Software-driven control of stereotaxic movement
- One micron resolution
- Download coordinates to computer for recall and/or archiving
- Angle adjustments
- Variable speed on 2-axis (dorsal-ventral)
- Digital Atlas integration
- Frame representation
- Setting of logical coordinate system (Bregma)
- Coordinates and Atlas visualization of the probe
- Advanced 3D visualization options
- Spatial Atlas representation
- Intuitive probe control
- Intuitive navigation
Motorized images:
- Axial view
- Coronal view
- Sagittal view
- Working view
Image manipulations include:
- Spatial Rotation
- Fixed Rotation
- Zooming
- Panning
Choice of two options:
- Add-on Motorized Manipulator Arm and Axis StereoDrive Software
Keep existing arm and have a new one shipped - Conversion to Motorized Manipulator Arm and Axis StereoDrive Software
Send existing arm for conversion to motorized
Specifications
Neuroscience | |
3 axes | |
Mouse, Rat |