Learn More
World Precision Instrument Blumenthal Rongeurs, 15cm, German
Supplier: World Precision Instrument 14091G
Blumenthal Rongeurs are used for removing bone or other hard tissue during surgical procedures. Blumenthal Rongeurs are designed with a pointed, triangular cutting tip designed for precise cutting and shaping of bone and other hard tissue. They can be used in a variety of procedures, including orthopedic surgeries, neurosurgery, and plastic surgery. They may be used in studies examining bone growth or regeneration, or in studies exploring the effects of various drugs or treatments on bone tissue.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.