Learn More
STEMCELL Technologies RosetteSep™ Human Multiple Myeloma Cell Enrichment Cocktail, For processing: 200 mL bone marrow
Supplier: STEMCELL Technologies 15169
The RosetteSep™ Human Multiple Myeloma Cell Enrichment Cocktail is designed to enrich multiple myeloma cells (B cell and plasma cells) from fresh bone marrow aspirates by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD2, CD14, CD33, CD41, CD45RA, CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs. The purified multiple myeloma cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium. Products are for Research Use Only, Not for Diagnostic or Therapeutic use, and are not to be administered to humans.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.