Learn More
STEMCELL Technologies RosetteSep Human Multiple Myeloma Cell Enrichment Cocktail, For processing: 40 mL bone marrow processed
SDP
Supplier:  STEMCELL Technologies 15129
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
The RosetteSep Human Multiple Myeloma Cell Enrichment Cocktail is designed to enrich multiple myeloma cells (B cell and plasma cells) from fresh bone marrow aspirates by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD2 CD14 CD33 CD41 CD45RA CD66b and glycophorin A on red blood cells (RBCs). When centrifuged over a buoyant density medium such as Lymphoprep (Catalog 07801) the unwanted cells pellet along with the RBCs. The purified multiple myeloma cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium.
Catalog No. 50-239-0396
missing translation for 'QuantityLeft'