Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rosuvastatin calcium
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Rosuvastatin calcium is a potent HMG-CoA reductase inhibitor (IC50 = 5.4 nM). Inhibits cholesterol synthesis in rat hepatocytes in vitro and in vivo. Reduces plasma LDL cholesterol levels. Enhances lentiviral transduction of natural killer cells. Orally bioavailable.
Specifications
Specifications
Chemical Name or Material | (3 R,5 S,6 E)-7-[4-(4-Fluorophenyl)-6-isopropyl-2-[methyl(methylsulfonyl)amino]pyrimidin-5-yl])-3,5-dihydroxyhept-6-enoic acid hemicalcium salt |
CAS | 147098-20-2 |
Assay Percent Range | >99% |
Quantity | 10 mg |
Target | HMG-CoA Reductase Inhibitors |
Molecular Formula | C22H27FN3O6S.1⁄2Ca |
Formula Weight | Observed MW: 500.57 |
Purity | 99% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction