Learn More
Hettich Centrifuges ROTANTA 460,(12) Plate, BioPackage
SDP
Supplier:  Hettich Centrifuges 460PLATEHSBC
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Hettich ROTANTA 460 PLATE PACKAGE HIGH-SPEED BIO-CONTAINMENT, (1) Centrifuge, (1) Horizontal Swing-out Rotor, (2) Buckets, (2) Adapters, (2) Bio-containment lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7570
missing translation for 'QuantityLeft'