Learn More
Hettich Centrifuges ROTANTA 460RF, (24)Plate,BCPackage
SDP
Supplier:  Hettich Centrifuges 460RFPLATEBC
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Hettich ROTANTA 460 RF PLATE PACKAGE BIO-CONTAINMENT, (1) Centrifuge, (1) Horizontal Swing-out Rotor, (4) Buckets, (4) Adapters, (4) Bio-containment lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7542