Learn More
Hettich Centrifuges ROTANTA460R, (24)Plate, BioPackage
SDP
Supplier:  Hettich Centrifuges 460RPLATEBC
Encompass_Preferred
Hettich ROTANTA 460 R PLATE PACKAGE BIO-CONTAINMENT, (1) Centrifuge, (1) Horizontal Swing-out Rotor, (4) Buckets, (4) Adapters, (4) Bio-containment lids, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7539
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.